missing translation for 'onlineSavingsMsg'
Learn More

Warfarin, TRC

Product Code. 17937399
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17937399 2.5 g 2.5g
17997839 1 g 1g
17999329 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17937399 Supplier Toronto Research Chemicals Supplier No. TRCW4985002.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-81-2
Molecular Formula C19 H16 O4
Molecular Weight (g/mol) 308.33
Synonym 2H-1-Benzopyran-2-one, 4-hydroxy-3-(3-oxo-1-phenylbutyl)-, Coumarin, 3-(α-acetonylbenzyl)-4-hydroxy- (7CI,8CI), 4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one, (RS)-Warfarin, (±)-Warfarin, (±)-Warfarin-alcohol, 1-(4'-Hydroxy-3'-coumarinyl)-1-phenyl-3-butanone, 3-(1'-Phenyl-2'-acetylethyl)-4-hydroxycoumarin, 3-(α-Acetonylbenzyl)-4-hydroxycoumarin, 3-(α-Phenyl-β-acetylethyl)-4-hydroxycoumarin, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2H-chromen-2-one, Athrombine-K, Brumolin, Circuvit, Co-Rax, Compound 42, Coumafen, Coumafene, Coumaphen, Coumefene, DL-3-(α-Acetonylbenzyl)-4-hydroxycoumarin, Dethmor, Kumader, Kumadu, Kumatox, Martefarin, NSC 59813, Ratron, Ratron G, Rodafarin, Rodafarin C, Rodex, Temus W, Vampirinip II, Vampirinip III, W.A.R.F. 42, WARF compound 42, Warf 5, Warfarin, Warfarin acid, Zoocoumarin, rac-Warfarin
IUPAC Name 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one
SMILES CC(=O)CC(C1=C(O)c2ccccc2OC1=O)c3ccccc3

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,21H,11H2,1H3
Formula Weight 308.1049
Percent Purity >95%
Chemical Name or Material Warfarin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.