missing translation for 'onlineSavingsMsg'
Learn More

Water Pollution QC Check Sample: Complex Nutrients

Product Code. 30316772
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316772 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316772 Supplier VHG Labs Supplier No. VHGQWPCNUT15
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphorus∣Total Kjeldahl Nitrogen
CAS 7723-14-0
Analyte or Component Names Phosphorus∣Total Kjeldahl Nitrogen
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Dilute HCl
Industry Type Petrochemical Testing
Quantity 15 mL
Concentration or Composition (by Analyte or Components) Total Kjeldahl Nitrogen 3.78 mg/L∣Phosphorus 17.5 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.