missing translation for 'onlineSavingsMsg'
Learn More

Water Pollution QC Check Sample: Demand

Product Code. 30316408
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30316408 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316408 Supplier VHG Labs Supplier No. VHGQWPDEM15
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Biochemical Oxygen Demand (BOD)∣Carbonaceous BOD (CBOD)∣Chemical Oxygen Demand (COD)∣Total Organic Carbon
Analyte or Component Names Biochemical Oxygen Demand (BOD)∣Carbonaceous BOD (CBOD)∣Chemical Oxygen Demand (COD)∣Total Organic Carbon
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Dilute HCl
Industry Type Petrochemical Testing
Quantity 15 mL
Concentration or Composition (by Analyte or Components) Total Organic Carbon 79.8 mg/L∣Carbonaceous BOD (CBOD) 112 mg/L∣Biochemical Oxygen Demand (BOD) 124 mg/L∣Chemical Oxygen Demand (COD) 202 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.