missing translation for 'onlineSavingsMsg'
Learn More

Water Pollution QC Check Sample: Hardness

Product Code. 30325876
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325876 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325876 Supplier VHG Labs Supplier No. VHGQWPHRD500
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium∣Calcium hardness∣Magnesium∣Non-Filterable Residue∣Total hardness
CAS 7440-70-2, 7439-95-4
Analyte or Component Names Calcium∣Calcium hardness∣Magnesium∣Non-Filterable Residue∣Total hardness
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix Water
Industry Type Petrochemical Testing
Quantity 1 Ea.
Concentration or Composition (by Analyte or Components) Magnesium 24.8 mg/L∣Non-Filterable Residue 80.6 mg/L∣Calcium 82.6 mg/L∣Calcium hardness 206 mg/L∣Total hardness 308 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.