missing translation for 'onlineSavingsMsg'
Learn More

Water Pollution QC Check Sample: Oil and Grease Concentrate

Product Code. 30315154
Change view
Click to view available options
Quantity:
23 mL
Unit Size:
23mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315154 23 mL 23mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315154 Supplier VHG Labs Supplier No. VHGQWPOG23
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oil and Grease
Analyte or Component Names Oil and Grease
Shipping Condition Room Temperature
Concentration 96.5 mg/L
Solution Type Mixture
IUPAC Name Oil and Grease
Solvent or Matrix n-Propanol
Industry Type Petrochemical Testing
Quantity 23 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.