missing translation for 'onlineSavingsMsg'
Learn More

Water Pollution QC Check Sample: Simple Nutrients

Product Code. 30323278
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323278 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323278 Supplier VHG Labs Supplier No. VHGQWPSNUT15

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonia∣Nitrate∣Nitrite∣Ortho-phosphate
CAS 7664-41-7, 14797-55-8, 14797-65-0, 7558-79-4
Analyte or Component Names Ammonia∣Nitrate∣Nitrite∣Ortho-phosphate
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Dilute HCl
Industry Type Petrochemical Testing
Quantity 15 mL
Concentration or Composition (by Analyte or Components) Ortho-phosphate 2.4 mg/L∣Ammonia 10.8 mg/L∣Nitrate 16.1 mg/L∣Nitrite 16.1 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.