missing translation for 'onlineSavingsMsg'
Learn More

Water Pollution QC Check Sample: Trace Metals

Product Code. 30312024
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312024 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312024 Supplier VHG Labs Supplier No. VHGQWPTM15
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium∣Antimony∣Arsenic∣Barium∣Beryllium∣Boron∣Cadmium∣Chromium∣Cobalt∣Copper∣Iron∣Lead∣Manganese∣Molybdenum∣Nickel∣Selenium∣Silver∣Strontium∣Thallium∣Vanadium∣Zinc
CAS 7429-90-5, 7440-36-0, 7440-38-2, 7440-39-3, 7440-41-7, 7440-42-8
Analyte or Component Names Aluminium∣Antimony∣Arsenic∣Barium∣Beryllium∣Boron∣Cadmium∣Chromium∣Cobalt∣Copper∣Iron∣Lead∣Manganese∣Molybdenum∣Nickel∣Selenium∣Silver∣Strontium∣Thallium∣Vanadium∣Zinc
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Quantity 15 mL
Concentration or Composition (by Analyte or Components) Manganese 767193 μg/L∣Zinc 100-2000 μg/L∣Barium 100-2500 μg/L∣Chromium 17-1000 μg/L∣Aluminium 200-4000 μg/L∣Iron 200-4000 μg/L∣Silver 26-600 μg/L∣Cobalt 28-1000 μg/L∣Strontium 30-300 μg/L∣Copper 40-900 μg/L∣Vanadium 55-2000 μg/L∣Molybdenum 60-600 μg/L∣Thallium 60-900 μg/L∣Lead 70-3000 μg/L∣Arsenic 70-900 μg/L∣Boron 800-2000 μg/L∣Nickel 80-3000 μg/L∣Cadmium 8-750 μg/L∣Beryllium 8-900 μg/L∣Selenium 90-2000 μg/L∣Antimony 95-900 μg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.