missing translation for 'onlineSavingsMsg'
Learn More

Water Supply QC Check Sample: Organic Carbon

Product Code. 30317096
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317096 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317096 Supplier VHG Labs Supplier No. VHGQWSOC15
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dissolved Organic Carbon∣Total Organic Carbon
Analyte or Component Names Dissolved Organic Carbon∣Total Organic Carbon
Shipping Condition Room Temperature
Solution Type Single Solution
Solvent or Matrix Dilute H2SO4
Industry Type Petrochemical Testing
Quantity 15 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.