missing translation for 'onlineSavingsMsg'
Learn More

Water Supply QC Check Sample: Trace Metals

Product Code. 30325649
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325649 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325649 Supplier VHG Labs Supplier No. VHGQWSTM15
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium∣Antimony∣Arsenic∣Barium∣Beryllium∣Boron∣Cadmium∣Chromium∣Copper∣Iron∣Lead∣Manganese∣Molybdenum∣Nickel∣Selenium∣Silver∣Thallium∣Vanadium∣Zinc
CAS 7429-90-5, 7440-36-0, 7440-38-2, 7440-39-3, 7440-41-7, 7440-42-8
Analyte or Component Names Aluminium∣Antimony∣Arsenic∣Barium∣Beryllium∣Boron∣Cadmium∣Chromium∣Copper∣Iron∣Lead∣Manganese∣Molybdenum∣Nickel∣Selenium∣Silver∣Thallium∣Vanadium∣Zinc
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Quantity 15 mL
Concentration or Composition (by Analyte or Components) Cadmium 18295 μg/L∣Arsenic 18384 μg/L∣Antimony 18415 μg/L∣Beryllium 45931 μg/L∣Thallium 45932 μg/L∣Iron 100-1800 μg/L∣Selenium 10-100 μg/L∣Chromium 10-200 μg/L∣Nickel 10-500 μg/L∣Aluminium 130-2500 μg/L∣Molybdenum 15-130 μg/L∣Silver 20-300 μg/L∣Vanadium 315-2500 μg/L∣Zinc 400-2500 μg/L∣Manganese 40-900 μg/L∣Barium 500-3000 μg/L∣Copper 50-2000 μg/L∣Lead 5-100 μg/L∣Boron 800-2000 μg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.