missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Waymouth's Medium

Product Code. 11574426 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
5000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11574426 500 mL 500mL
11584426 10 x 500 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11574426 Supplier Gibco™ Supplier No. 31220023
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Waymouth's Medium

Waymouth's Medium is a chemically defined medium developed by Charity Waymouth and originally formulated for studies on the nutrition, metabolism, and growth of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Gibco™ Waymouth's Medium is also used for the culture of whole organs and a broad range of cell types, such as carcinoma cell lines.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.

Dual-Site cGMP Manufacturing
Gibco™ Waymouth's Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Waymouth's Medium product made in our Grand Island facility (11220-035). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Waymouth's Medium contains no proteins, lipids, or growth factors. Some cell types may required the supplementation with Fetal Bovine Serum (FBS). Waymouth's Medium uses a sodium bicarbonate buffer system (2.24 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Waymouth's Medium
With Additives Glutamine, Phenol Red
Without Additives No HEPES
For Use With (Application) Mammalian cell culture
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
Regulatory Status In-vitro Diagnostic Use
Content And Storage 2 to 8°C
Description Waymouth's Medium, 500mL, Liquid Form, With L-glutamine, Phenol Red, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.