missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Waymouth's Medium

Description
Waymouth's Medium is a chemically defined medium developed by Charity Waymouth and originally formulated for studies on the nutrition, metabolism, and growth of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Gibco™ Waymouth's Medium is also used for the culture of whole organs and a broad range of cell types, such as carcinoma cell lines.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.
Dual-Site cGMP Manufacturing
Gibco™ Waymouth's Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Waymouth's Medium product made in our Grand Island facility (11220-035). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Waymouth's Medium contains no proteins, lipids, or growth factors. Some cell types may required the supplementation with Fetal Bovine Serum (FBS). Waymouth's Medium uses a sodium bicarbonate buffer system (2.24 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.
Dual-Site cGMP Manufacturing
Gibco™ Waymouth's Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Waymouth's Medium product made in our Grand Island facility (11220-035). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Waymouth's Medium contains no proteins, lipids, or growth factors. Some cell types may required the supplementation with Fetal Bovine Serum (FBS). Waymouth's Medium uses a sodium bicarbonate buffer system (2.24 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
Specifications
| Form | Liquid |
| Product Type | Waymouth's Medium |
| With Additives | Glutamine, Phenol Red |
| Without Additives | No HEPES |
| For Use With (Application) | Mammalian cell culture |
| Quantity | 10 x 500 mL |
| Certifications/Compliance | ISO 13485 standards, FDA |
| Regulatory Status | For in-vitro diagnostic use |
| Content And Storage | 2 to 8°C |
| Description | Waymouth's Medium, 500mL, Liquid Form, With L-glutamine, Phenol Red, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?