missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBSCR17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
386.00 € - 529.00 €
Specifications
| Antigen | WBSCR17 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18479890
|
Novus Biologicals
NBP1-81843-25ul |
25 μL |
386.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18259825
|
Novus Biologicals
NBP1-81843 |
0.1 mL |
529.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WBSCR17 Polyclonal specifically detects WBSCR17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| WBSCR17 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 64409 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RPIAVRSGDAFHEIRPRAEVANLSAHSASPIQDAVLKRLSLLEDIVYRQLNGLSKSLGLIEGYGGRGKGGLPATLSPAEEEKAKGPHEKYGYNSYLSE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| DKFZp434I2216, EC 2.4.1.41, GalNAc-T-like protein 3, GALNT16, GALNT20, GALNTL3DKFZp761D2324, Polypeptide GalNAc transferase-like protein 3, polypeptide N-acetylgalactosaminyltransferase WBSCR17, pp-GalNAc-transferase 20, pp-GaNTase-like protein 3, Protein-UDP acetylgalactosaminyltransferase-like protein 3, putative polypeptide N-acetylgalactosaminyltransferase-like protein 3, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like 3, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 3, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 16, Williams-Beuren syndrome chromosomal region 17 protein, Williams-Beuren syndrome chromosome region 17 | |
| WBSCR17 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto