missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDHD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21217-25ul
This item is not returnable.
View return policy
Description
WDHD1 Polyclonal antibody specifically detects WDHD1 in Human samples. It is validated for Western Blot
Specifications
| WDHD1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml | |
| Acidic nucleoplasmic DNA-binding protein 1, AND1, AND-1, CHTF4, CTF4, CTF4, chromosome transmission fidelity factor 4 homolog, WD repeat and HMG-box DNA binding protein 1, WD repeat and HMG-box DNA-binding protein 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QTLNIVTWSPCGQYLAAGSINGLIIVWNVETKDCMERVKHEKGYAICGLAWHPTCGRISYTDAEGNLGLLENVCDPSGKTSSSKVSSRVEKDYNDLFDGDDMSNAGDFLNDNAVE | |
| 25 μg | |
| Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
| 11169 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction