missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR11 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | WDR11 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620671
|
Novus Biologicals
NBP2-93248-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625940
|
Novus Biologicals
NBP2-93248-0.1ml |
0.1 mL |
470.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WDR11 Polyclonal antibody specifically detects WDR11 in Human samples. It is validated for Western BlotSpecifications
| WDR11 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 55717 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Bromodomain and WD repeat-containing protein 2, BRWD2DKFZp434L1715, DR11, FLJ10506, FLJ42531, KIAA1351bromodomain and WD repeat domain containing 2, WD repeat domain 11, WD repeat domain 15, WD repeat-containing protein 11, WD repeat-containing protein 15, WDR15 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 975-1224 of human WDR11 (NP_060587.8). YFQKFQLERVNLQEVKRSTYDHTRKCTDQLLLLGQTDRAVQLLLETSADNQHYYCDSLKACLVTTVTSSGPSQSTIKLVATNMIANGKLAEGVQLLCLIDKAADACRYLQTYGEWNRAAWLAKVRLNPEECADVLRRWVDHLCSPQVNQKSKALLVLLSLGCFFSVAETLHSMRYFDRAALFVEACLKYGAFEVTEDTEKLITAIYADYARSLKNLGFKQGAVLFASKAGAAGKDLLNELESPKEEPIEE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Individuate un'opportunità di miglioramento?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title