missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDR25 Recombinant Mouse Monoclonal Antibody (K9N019_16F2)
GREENER_CHOICE

Product Code. 30829257
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829257 100 μL
30830345 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829257 Supplier Invitrogen™ Supplier No. MA564458

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: EAETEHAGSF NATGQQKDTS GVARPPGQDF ASGTLDVPKA GAQPTKHGSC EDPGGYRLPL AQLGRSDWGS CPSQRLQWPG KEPQVTFPIK EPSCSSLWTS HVPASHMPLA AARFKQVKLS RNFPKSSFHA QSESETVGKN GSSFQKKKCE DCVVPYTPRR LRQRQALSTE TGKGKDVEPQ Highest antigen sequence identity to the following orthologs: Mouse - 57%, Pig - 58%, Cynomolgus monkey - 89%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR25
Applications Immunoprecipitation
Classification Recombinant Monoclonal
Clone K9N019_16F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene WDR25
Gene Accession No. Q64LD2
Gene Alias C14orf67; WD repeat domain 25; WD repeat-containing protein 25; WDR25
Gene Symbols WDR25
Host Species Mouse
Immunogen Recombinant human WDR25 (aa 21-200).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79446
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.