missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR60 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
292.00 € - 539.00 €
Specifications
| Antigen | WDR60 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockout Validated, Knockdown Validated |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18456841
|
Novus Biologicals
NBP1-90437-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18244597
|
Novus Biologicals
NBP1-90437 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WDR60 Polyclonal specifically detects WDR60 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| WDR60 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ10300, FLJ23575, WD repeat domain 60, WD repeat-containing protein 60 | |
| WDR60 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockout Validated, Knockdown Validated | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 55112 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DIQTEEIETREVWTQHPGESTVVSGGSEQRDTSDAVVMPKIDTPRLCSFLRAACQVMAVLLEEDRLAAEPSWNLRAQDRALYFSDSSSQLNTSLPFLQNRKVSSLHTSRVQRQMVVSVH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title