missing translation for 'onlineSavingsMsg'
Learn More

WEHI-539 hydrochloride, MedChemExpress

Product Code. 16496678
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496678

Brand: Medchem Express HY15607A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WEHI-539 hydrochloride is a selective inhibitor of Bcl-XL with an IC50 of 1.1 nM.

WEHI-539 hydrochloride is a selective inhibitor of Bcl-XL with an IC50 of 1.1 nM.

Specifications

Chemical Name or Material WEHI-539 hydrochloride
CAS 2070018-33-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H30ClN5O3S2
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (80.62 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES OC(C1=C(CCCOC2=CC=C(CN)C=C2)SC(C3=CC4=C(CCC/C4=N\NC5=NC(C=CC=C6)=C6S5)C=C3)=N1)=O.[H]Cl
Molecular Weight (g/mol) 620.18
Formula Weight 620.18
Percent Purity 95.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.