missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-(1,3-oxazol-5-yl)benzoate, 97%, Thermo Scientific™

Product Code. 10001434
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10001434 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10001434 Supplier Thermo Scientific Maybridge Supplier No. CC14526DA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 898288-91-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C16H6F5NO3
MDL Number MFCD09025822
Quantity 1 g
Synonym pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, perfluorophenyl 4-oxazol-5-yl benzoate, 4-5-oxazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester, benzoic acid,4-5-oxazolyl-, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentakis fluoranyl phenyl 4-1,3-oxazol-5-yl benzoate
InChI Key FWUPBFHGEDUEID-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C2=CN=CO2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 4-(1,3-oxazol-5-yl)benzoate
Molecular Weight (g/mol) 355.22
PubChem CID 18525706
Formula Weight 355.22
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 4-(1, 3-oxazol-5-yl)benzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.