Learn More
Thermo Scientific™ Well Plate Sealing Mat Cutter
Product Code. 32226646 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
32226646 1 Ea. Each
1 options
Product Code. 32226646 Supplier Thermo Scientific™ Supplier No. 60180M975
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Effectively seal all microplate formats to minimize evaporation and protect samples from contamination and spilling.
Effectively seal all microplate formats to minimize evaporation and protect samples from contamination and spilling, ensure a consistent, reliable seal with all microplate formats with this easy-to-use, handheld applicator, and get significantly quicker dry-down times than standard methods, and remove most chromatography solvents in just minutes with easy to use 96-well sample concentrators.
WebSeal Cutter
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
Product Type Sealing Mat Cutter
Product Line Webseal