Learn More
Invitrogen™ West Nile Virus Envelope (lineage 1, strain NY99) Recombinant Rabbit Monoclonal Antibody (087)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560367
 View more versions of this product
Product Code. 30535136
  • 429.00 € / 100µL
Description
West Nile Virus Envelope (lineage 1, strain NY99) Recombinant Monoclonal Antibody for Western Blot
West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins which play a major role for WNV entry into target cells. The viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway as delivery of core gene delivery into the striatum of mouse brain and skeletal muscle resulted in cell death and inflammation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
West Nile Virus Envelope (lineage 1, strain NY99)
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
Western Blot
087
Unconjugated
West Nile envelope; West Nile Virus; WNV envelope
Recombinant West Nile Virus (WNV) (lineage 1, strain NY99) E/Envelope Protein (Domain III) protein (Gln586-Ala705).
100 μL
Primary
-20°C, Avoid Freeze/Thaw Cycles