missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ 96-Well Lysate Clarification Plate

Product Code. 11917137
Change view
Click to view available options
Pore Size:
0.45 μM
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
11917137 0.45 μM Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11917137 Supplier Cytiva Supplier No. 77700062

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for the removal of cellular debris from lysates during high-throughput plasmid DNA purification

  • Polystyrene plate with a proprietary filter membrane
  • Processes 96 lysates in <10 min.
  • Compatible with vacuum filtration or centrifugation
  • Produces recovery rates 10 to 30% higher than manual centrifuge methods
  • Recovers an average of 6μg DNA/well when used with the Whatman DNA Binding Plate (Cat. No. PF77002810)
  • Adaptable to automated platforms

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Description UNIFILTER 96-well microplate, 800 μl, long drip director
Array 8 x 12
Pore Size 0.45 μM
Color Clear
Material Polystyrene
Volume (Metric) Well 800 μL
No. of Wells 96
Well Shape Filter
For Use With (Application) An important tool for high-throughput plasmid DNA purification
Includes Long drip director
Quantity 25/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.