missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Fast Flow Protein Precipitation Microplate

Product Code. 11318714
Change view
Click to view available options
No. of Wells:
96
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells unitSize
11318714 96 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11318714 Supplier Cytiva Supplier No. 77207236

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Fast, easy and automatable protein precipitation

  • Prepare 96 samples up to 150μL for plasma and 200μL for serum in less than 10 minutes
  • Minimal liquid handling—precipitation and filtration performed in same well
  • Utilizes acetonitrile precipitation and vacuum filtration to remove protein contaminants from samples before downstream analysis
  • Oleophobic membrane configuration retains acetonitrile/sample mixture during mixing
  • Vacuum filtration allows mixture to pass through the membrane while precipitated proteins remain in well
  • Filtrate is then ready for HPLC injection or target UV quantitation

Plasma monitoring for pharmaceutical efficacy and new drug development

TRUSTED_SUSTAINABILITY

Especificaciones

Format Standard
Description GE’s Whatman UNIFILTER 2 mL, 96 well microplates are used for large sample or reagent volumes, as in biomolecule purification by SPE and organic synthesis in combinatorial chemistry library generation.
Array 8 x 12
Product Type UNIFILTER Microplate, 96-well, 2 mL, protein precipitation fast flow, filter bottom with long drip d
Material Glass-filled Polypropylene
Volume (Metric) Well 2000 μL
No. of Wells 96
Well Shape Filter
For Use With (Application) Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation
Includes Long drip director
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.