missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Stainless Steel Support Screens

Product Code. 11768959 Shop All Wheaton Science Products Products
Change view
Click to view available options
For Use With (Equipment):
25mm Vacuum Filtration Assembly
47mm Vacuum Filtration Assembly
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11768959 25mm Vacuum Filtration Assembly Each
11591315 47mm Vacuum Filtration Assembly Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11768959 Supplier DWK Life Sciences Supplier No. 419337

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum filtration assembly. Wheaton™ Stainless Steel Support Screens are manufactured from the highest quality materials.

  • Stainless Steel
  • For use with 25mm assembly
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 25mm Vacuum Filtration Assembly
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.