missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ WI-38 Whole Cell Lysate

Product Code. 18038387 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18038387 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18038387 Supplier Novus Biologicals™ Supplier No. NBP2102650.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

WI-38 (Human Lung Fibroblast, normal) whole respirator cell lysate 2mg/mL supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. Growth media: ATCC medium. Lysates are guaranteed for 6 months from date of receipt.

The WI-38 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Spezifikation

For Use With (Application) Western Blot
Tissue WI-38
Quantity 0.1 mg
Storage Buffer Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol.
Test Specificity WI-38 (Human Lung Fibroblast, normal)
Host Species Human
Concentration 2 mg/mL
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.