missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA512299
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse, rat, Xenopus and zebrafish based on sequence homology.
The protein encoded by this gene functions to inhibit WNT proteins, which are extracellular signaling molecules that play a role in embryonic development. This protein contains a WNT inhibitory factor (WIF) domain and five epidermal growth factor (EGF)-like domains, and is thought to be involved in mesoderm segmentation.
Specifications
| WIF1 | |
| Polyclonal | |
| Unconjugated | |
| WIF1 | |
| AW107799; UNQ191/PRO217; WIF; WIF1; WIF-1; Wnt inhibitory factor 1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 11197 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q9Y5W5 | |
| WIF1 | |
| KLH conjugated synthetic peptide between 51-80 amino acids from the N-terminal region of human WIF1 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction