missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ William's E Medium, GlutaMAX™ Supplement
GREENER_CHOICE

Product Code. 11514466
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11514466 10 x 500 mL Pack of 10
10043282 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11514466 Supplier Gibco™ Supplier No. 32551087
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

William's E Medium was originally developed by Williams and Gunn as reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells. Gibco™ William's E Medium can also be used for growing hepatocyte cells of different species, such as human derived HepaRG™ cells.

This William's E Medium is modified as follows:

  • With: Phenol Red, GlutaMAX™
  • Without: HEPES

The complete formulation is available.

Gibco™ William's E Medium is a modification of earlier media that has been enriched in amino acids and double the glucose. William's E Medium contains unique ingredients, including zinc, iron, manganese, non-essential amino acids, the reducing agent glutathione and the lipid methyl linoleate. Gibco™ William's E Medium with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

cGMP Manufacturing

Gibco™ William's E Medium is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

William's E Medium contains no proteins or growth factors. Therefore, William's E Medium requires supplementation, commonly with 5-10% Fetal Bovine Serum (FBS). William's E Medium uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Classification Protein-free, Animal Origin-free, Serum-free
Form Liquid
Product Type William's E Medium
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives GlutaMAX, Phenol Red
Without Additives No HEPES
Product Line GlutaMAX
Quantity 10 x 500 mL
Content And Storage Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Culture Type Mammalian Cell Culture
Show More Show Less
Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?

Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.

Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?

In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.