missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wilm's Tumor 1 Rabbit, Clone: WT1/1434R, enQuireBio™
Rabbit Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | Wilm's Tumor 1 |
|---|---|
| Clone | WT1/1434R |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15911098
|
EnquireBio
7490-RBM4-P0 |
20 μg | |||||||
|
15921098
|
EnquireBio
7490-RBM4-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Wilm's Tumor 1 | |
| 200μg/mL | |
| Monoclonal | |
| Rabbit | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| 7490 | |
| Recombinant human full-length WT1 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 47-55kDa |
| WT1/1434R | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_001185480 | |
| WT1 | |
| IgG κ | |
| Protein A/G Purified | |
| Nuclear and Cytoplasmic | |
| K562 cells. Wilm s Tumor, mesothelioma or fetal kidney. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title