missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIRE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
275.00 € - 528.00 €
Specifications
| Antigen | WIRE |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18444541
|
Novus Biologicals
NBP1-86856-25ul |
25 μL |
275.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18208846
|
Novus Biologicals
NBP1-86856 |
0.1 mL |
528.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WIRE Polyclonal specifically detects WIRE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| WIRE | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| PP10631, WAS/WASL interacting protein family, member 2, WIP-related protein | |
| WIPF2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 147179 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PPPYRMHGSEPPSRGKPPPPPSRTPAGPPPPPPPPLRNGHRDSITTVRSFLDDFESKYSFHPVEDFPAPEEYKHFQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title