missing translation for 'onlineSavingsMsg'
Learn More

Human Wnt-7a Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7112378
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18746988 25 μg 25µg
18706779 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18746988 Supplier R&D Systems Supplier No. MAB30081SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Wnt-7a Monoclonal specifically detects Wnt-7a in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wnt-7a
Applications Immunocytochemistry
Classification Monoclonal
Clone 946744
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O00755
Gene Alias protein Wnt-7a, proto-oncogene Wnt7a protein, wingless-type MMTV integration site family, member 7A, Wnt7a
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Wnt-7a Met1-Lys349 Accession # O00755
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7476
Test Specificity Detects human Wnt-7a in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.