missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT9B Monoclonal Antibody (964742)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561335
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
WNT9B, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
Specifications
| WNT9B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O35468 | |
| Wnt9b | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Wnt-9b, Met1-Arg359. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Neutralization | |
| 964742 | |
| Unconjugated | |
| Wnt9b | |
| clf; clf1; protein Wnt-14b; Protein Wnt-15; Protein Wnt-9b; UNQ6973/PRO21956; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9B; wingless-type MMTV integration site family member 9B; wingless-type MMTV integration site family, member 15; wingless-type MMTV integration site family, member 9B; Wnt family member 9B; Wnt14b; wnt-14b; Wnt15; wnt-15; Wnt9b | |
| Rat | |
| Protein A/G | |
| RUO | |
| 22412 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction