missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress WST-8
Click to view available options
Quantity:
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
25mg
50mg
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Product Type | Dye Reagents |
| Quantity | 25 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 83.33 mg/mL (138.54 mM; Need ultrasonic) ∣ DMSO : 10 mg/mL (16.63 mM; Need ultrasonic) |
| Target | HBV |
| Molecular Weight (g/mol) | 601.48 g/mol |
| CAS | 193149-74-5 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction