missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | WT1 |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16850414
|
Proteintech
12609-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16840414
|
Proteintech
12609-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Wilm′s Tumor (WT), a sporadic and familial childhood kidney tumor, is genetically heterogeneous. Wilm′s tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelioma.Specifications
| WT1 | |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| P19544, P22561, P49952 | |
| 22431, 24883, 7490 | |
| WT1 Fusion Protein Ag3337 | |
| Primary | |
| -20°C | |
| WT1 |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT 2, WT1, WT33 | |
| WT1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title