Learn More
WWOX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
363.00 € - 547.00 €
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18778943
View Documents
Novus Biologicals
NBP2-47579
547.00 €
0.10mL
View alternative products
Save Up to 
»
18615935
View Documents
Novus Biologicals
NBP2-47579-25ul
25 μL
363.00 €
25µL
View alternative products
Save Up to 
»
Description
Description
WWOX Polyclonal specifically detects WWOX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.