missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XAGE2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46807-25ul
This item is not returnable.
View return policy
Description
XAGE2 Polyclonal antibody specifically detects XAGE2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| XAGE2 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Q96GT9 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9502 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Cancer/testis antigen 12.2, cancer/testis antigen family 12, member 2, CT12.2GAGED3, G antigen family D member 3, G antigen, family D, 3, Protein XAGE-2, X antigen family, member 2, XAGE-2, XAGE2B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PTPDQKREDDQGAAEIQVPDLEADLQ | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction