missing translation for 'onlineSavingsMsg'
Learn More

Xanthene-9-carboxamide, TRC

Product Code. 30453769
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453769 500 mg 500mg
30453783 100 mg 100mg
30451652 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453769 Supplier Toronto Research Chemicals Supplier No. TRCX744003500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5813-90-1
Molecular Formula C14H11NO2
Molecular Weight (g/mol) 225.24
IUPAC Name 9H-xanthene-9-carboxamide
SMILES NC(=O)C1c2ccccc2Oc3ccccc13

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H11NO2/c15-14(16)13-9-5-1-3-7-11(9)17-12-8-4-2-6-10(12)13/h1-8,13H,(H2,15,16)
Formula Weight 225.079
Chemical Name or Material Xanthene-9-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.