missing translation for 'onlineSavingsMsg'
Learn More

XAV-939, MedChemExpress

Product Code. 16446646
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. Quantity unitSize
16446646 100 mg 100mg
16446328 10 mg 10mg
16441166 200 mg 200mg
16408917 5 mg 5mg
16402447 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16446646

Brand: Medchem Express HY15147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress XAV-939 is a potent tankyrase inhibitor that targets Wnt/β-catenin signaling. XAV-939 stabilizes axin by inhibiting tankyrase 1 and tankyrase 2 (IC50s of 5 and 2 nM, respectively), thereby stimulating β-catenin degradation. XAV939 binds tightly to the catalytic (PARP) domains of TNKS1 and TNKS2 (Kds of 99 and 93 nM, respectively).

XAV-939 is a potent tankyrase inhibitor that targets Wnt/β-catenin signaling. XAV-939 stabilizes axin by inhibiting tankyrase 1 and tankyrase 2 (IC50s of 5 and 2 nM, respectively), thereby stimulating β-catenin degradation. XAV939 binds tightly to the catalytic (PARP) domains of TNKS1 and TNKS2 (Kds of 99 and 93 nM, respectively).

Specifications

Chemical Name or Material XAV-939
Physical Form Solid
CAS 284028-89-3
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H11F3N2OS
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 25 mg/mL (80.05 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C(CSCC2)=C2N=C(C3=CC=C(C(F)(F)F)C=C3)N1
Molecular Weight (g/mol) 312.31
Formula Weight 312.31
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.