missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XBP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
244.00 € - 508.00 €
Specifications
| Antigen | XBP1 |
|---|---|
| Concentration | 0.58 mg/mL |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry reported in scientific literature (PMID 31031094), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100, KnockDown Validated reported in scientific literature (PMID 33513694) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18068314
|
Novus Biologicals
NBP1-77681SS |
0.025 mL |
244.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776663
|
Novus Biologicals
NBP1-77681 |
508.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
XBP1 Polyclonal specifically detects XBP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| XBP1 | |
| Western Blot 1 ug/mL, Flow Cytometry reported in scientific literature (PMID 31031094), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100, KnockDown Validated reported in scientific literature (PMID 33513694) | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| P17861, O35426 | |
| 7494 | |
| A genomic peptide made to an internal region of the human XBP1 protein (within residues 100-250). [Swiss-Prot P17861] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.58 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP1s, XBP1u, XBP2Tax-responsive element-binding protein 5 | |
| XBP1 | |
| IgG | |
| Affinity Purified | |
| This antibody is specific for both XBP1s and XBP1u. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title