Learn More
Novus Biologicals™ xCT Antibody (Agilvax Patent Anti-Slc7A11) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28188-50ug
 View more versions of this product
Product Code. 30048684
  • 213.00 € / 50µg
Description
xCT Monoclonal antibody specifically detects xCT in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
xCT
Monoclonal
Unconjugated
Q9UPY5
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Agilvax Patent Anti-Slc7A11
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, CCBR1, cystine/glutamate transporter, SLC7A11, solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11, Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCT
SLC7A11
50 μg
Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
23657
Human
Purified