missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPC Antibody (3.26), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 17 publications
Brand: Novus Biologicals NB100-477
This item is not returnable.
View return policy
Description
XPC Monoclonal specifically detects XPC in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| XPC | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin | |
| complementation group C p125, RAD4, Xeroderma pigmentosum group C-complementing protein, xeroderma pigmentosum, complementation group C, XP3, XPCCDNA repair protein complementing XP-C cells | |
| Mouse | |
| 123 kDa | |
| 100 μL | |
| Primary | |
| Mouse reactivity reported in scientific literature (PMID: 17229834). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3.26 | |
| Unconjugated | |
| Q01831 | |
| XPC | |
| Recombinant XPC purified from E. coli | |
| Protein G purified | |
| RUO | |
| 7508 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction