missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPO6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93480-0.1ml
This item is not returnable.
View return policy
Description
XPO6 Polyclonal antibody specifically detects XPO6 in Human, Mouse samples. It is validated for Western Blot
Specifications
| XPO6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Exp6, exportin 6, exportin-6, FLJ22519, KIAA0370EXP6, RAN binding protein 20, Ran-binding protein 20, RANBP20 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-530 of human XPO6 (NP_055986.1). ASVNGSSQNCVSGQERGRLGVLAMSCINELMSKNCVPMEFEEYLLRMFQQTFYLLQKITKDNNAHTVKSRLEELDESYIEKFTDFLRLFVSVHLRRIESYSQFPVVEFLTLLFKYTFHQPTHEGYFSCLDIWTLFLDYLTSKIKSRLGDKEAVLNRYEDALVLLLTEVLNRIQFRYNQAQLEELDDETLDDDQQTEWQRYLRQSLEVVAKVMELLPTHAFSTLFPVLQDNLEVYLGLQQFI | |
| 0.1 mL | |
| Signal Transduction | |
| 23214 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction