missing translation for 'onlineSavingsMsg'
Learn More

Xylenecyanol FF, Electrophoresis Reagent, dye content 70%

Product Code. 15430567
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 15430567

Brand: Thermo Scientific Alfa Aesar J66377.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2650-17-1 | C25H27N2NaO6S2 | 538.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2650-17-1
Molecular Formula C25H27N2NaO6S2
Molecular Weight (g/mol) 538.61
MDL Number MFCD00019481
InChI Key VVLFAAMTGMGYBS-KRQUPCAFSA-M
Synonym Acid Blue 147,C.I. 42135
IUPAC Name sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
SMILES [Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O

Specifications

Melting Point ∼295°C (decomposition)
Packaging Glass bottle
Quantity 10 g
Formula Weight 538.61
Percent Purity dye content 70%
Physical Form Powder
Chemical Name or Material Xylenecyanol FF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.