missing translation for 'onlineSavingsMsg'
Learn More

Y16, MedChemExpress

Product Code. 16445278
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16445278 10 mg 10mg
16407997 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16445278 Supplier Medchem Express Supplier No. HY12649

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Y16 is a specific inhibitor of Leukemia-associated Rho guanine nucleotide exchange factor (LARG) with a Kd value of 76 nM. Y16 is active in blocking the interaction of LARG and related G-protein-coupled Rho GEFs with RhoA. Y16 shows no detectable effect on other diffuse B-cell lymphoma (Dbl) family Rho GEFs, Rho effectors, or a RhoGAP.

Y16 is a specific inhibitor of Leukemia-associated Rho guanine nucleotide exchange factor (LARG) with a Kd value of 76 nM. Y16 is active in blocking the interaction of LARG and related G-protein-coupled Rho GEFs with RhoA. Y16 shows no detectable effect on other diffuse B-cell lymphoma (Dbl) family Rho GEFs, Rho effectors, or a RhoGAP.

Specifications

Chemical Name or Material Y16
CAS 429653-73-6
Color Red
Purity Grade Notes Research
Molecular Formula C24H20N2O3
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (65.03 mM; Need ultrasonic)
SMILES O=C(N1)/C(C(N1C2=CC=CC=C2)=O)=C\C3=CC(OCC4=CC(C)=CC=C4)=CC=C3
Molecular Weight (g/mol) 384.43
Formula Weight 384.43
Percent Purity 97.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.