missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-58358SS
This item is not returnable.
View return policy
Description
YAP1 Polyclonal specifically detects YAP1 in Human, Mouse, Rat, Canine, Zebrafish samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, Knockdown.
Specifications
| YAP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 15% glycerol with 0.02% Sodium Azide | |
| 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog | |
| Rabbit | |
| 48 kDa | |
| 0.025 mL | |
| Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases | |
| 10413 | |
| Human, Mouse, Rat, Canine, Zebrafish | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown | |
| 0.2 mg/mL | |
| Western Blot 1:1000, Simple Western 1:12.5, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-10 ug, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 28581498), Immunoblotting reported in scientific literature (PMID 28406163), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated reported in scientific literature (PMID 28406163) | |
| P46937 | |
| YAP1 | |
| This YAP1 Antibody was developed against a partial recombinant human YAP1 protein expressed in bacteria. [Uniprot: P46937], N-terminal GST fusion protein | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected reactivity based on immunogen homology: Isoform 4 (100%), Isoform 6 (100%) | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction