Learn More
YAP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB110-58358
 View more versions of this product
Product Code. 18785923
  • 607.00 € / 0.10mL
Description
YAP1 Polyclonal specifically detects YAP1 in Human, Mouse, Rat, Canine, Zebrafish samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, Knockdown.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Canine, Zebrafish
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.