Learn More
YPEL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41163
 View more versions of this product
Product Code. 18132059
  • 507.00 € / 0.10mg
Description
YPEL1 Polyclonal specifically detects YPEL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
YPEL1
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
YPEL1
Antibody was raised against a 11 amino acid synthetic peptide near the amino terminus of human YPEL1. The immunogen is located within the first 50 amino acids of YPEL1. Amino Acid Squence: AAVKMTKSKTFQA
0.1 mg
Primary
Human, Mouse, Rat
IgG
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5 μg/mL
DiGeorge syndrome-related protein, MGC64992, protein yippee-like 1, yippee (Drosophila) homolog-like 1, yippee-like 1 (Drosophila)
Rabbit
Affinity Purified
RUO
29799
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.