missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YPEL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41163-0.025mg
This item is not returnable.
View return policy
Description
YPEL1 Polyclonal specifically detects YPEL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| YPEL1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| YPEL1 | |
| Antibody was raised against a 11 amino acid synthetic peptide near the amino terminus of human YPEL1. The immunogen is located within the first 50 amino acids of YPEL1. Amino Acid Squence: AAVKMTKSKTFQA | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5 μg/mL | |
| DiGeorge syndrome-related protein, MGC64992, protein yippee-like 1, yippee (Drosophila) homolog-like 1, yippee-like 1 (Drosophila) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 29799 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction