Learn More
YPEL2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41164MFV610
Product Code. 18752743
  • 655.00 € / 0.10mL
Description
YPEL2 Polyclonal antibody specifically detects YPEL2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
YPEL2
Polyclonal
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
DiGeorge syndrome-related protein, Protein Yippee-Like 2, Yippee-Like 2 (Drosophila)
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human YPEL2. The immunogen is located within amino acids 30 - 80 of YPEL2. Amino Acid Squence: SNPMPQTGEVYPAPGS
0.1 mL
Cancer, Cell Cycle and Replication
388403
Store at 4C in the dark.
IgG
Western Blot, ELISA, Immunocytochemistry
mFluor Violet 610 SE
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse
Purified