missing translation for 'onlineSavingsMsg'
Learn More

YQ/T 31-2013 Photoinitiators Mixture 525 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30325475
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325475 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325475 Supplier Dr. Ehrenstorfer Supplier No. DREA50000525AL
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2-Methylphenyl)phenylmethanone (2-Methylbenzophenone)∣1-Benzoyl-1-hydroxycyclohexane∣2,2-Dimethoxy-2-phenylacetophenone∣2,4-Diethylthioxanthone∣2-Benzoylbenzoic acid methyl ester∣2-Hydroxy-2-methylpropiophenone∣2-Isopropylthioxantone ITX∣2-methyl-4'-(methylthio)-2-Morpholinopropiophenone∣3-Methylbenzophenone∣4,4-Bis(diethylamino)benzophenone∣4,4'-Bis(dimethylamino)benzophenone∣4-Dimethylaminobenzoic acid ethyl ester∣4-Isopropylthioxantone∣4-Methylbenzophenone∣4-Phenylbenzophenon∣Benzophenone∣Methyl 2-oxo-2-Phenylacetate (Methyl Benzoylformate)∣Padimate O
CAS 131-58-8, 947-19-3, 24650-42-8, 82799-44-8, 606-28-0, 7473-98-5
Analyte or Component Names (2-Methylphenyl)phenylmethanone (2-Methylbenzophenone)∣1-Benzoyl-1-hydroxycyclohexane∣2,2-Dimethoxy-2-phenylacetophenone∣2,4-Diethylthioxanthone∣2-Benzoylbenzoic acid methyl ester∣2-Hydroxy-2-methylpropiophenone∣2-Isopropylthioxantone ITX∣2-methyl-4'-(methylthio)-2-Morpholinopropiophenone∣3-Methylbenzophenone∣4,4-Bis(diethylamino)benzophenone∣4,4'-Bis(dimethylamino)benzophenone∣4-Dimethylaminobenzoic acid ethyl ester∣4-Isopropylthioxantone∣4-Methylbenzophenone∣4-Phenylbenzophenon∣Benzophenone∣Methyl 2-oxo-2-Phenylacetate (Methyl Benzoylformate)∣Padimate O
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (2-Methylphenyl)phenylmethanone (2-Methylbenzophenone) 100 μg/mL∣1-Benzoyl-1-hydroxycyclohexane 100 μg/mL∣2,2-Dimethoxy-2-phenylacetophenone 100 μg/mL∣2,4-Diethylthioxanthone 100 μg/mL∣2-Benzoylbenzoic acid methyl ester 100 μg/mL∣2-Hydroxy-2-methylpropiophenone 100 μg/mL∣2-Isopropylthioxantone ITX 100 μg/mL∣2-methyl-4'-(methylthio)-2-Morpholinopropiophenone 100 μg/mL∣3-Methylbenzophenone 100 μg/mL∣4,4-Bis(diethylamino)benzophenone 100 μg/mL∣4,4'-Bis(dimethylamino)benzophenone 100 μg/mL∣4-Dimethylaminobenzoic acid ethyl ester 100 μg/mL∣4-Isopropylthioxantone 100 μg/mL∣4-Methylbenzophenone 100 μg/mL∣4-Phenylbenzophenon 100 μg/mL∣Benzophenone 100 μg/mL∣Methyl 2-oxo-2-Phenylacetate (Methyl Benzoylformate) 100 μg/mL∣Padimate O 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.