missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YTHDC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
285.00 € - 500.00 €
Specifications
| Antigen | YTHDC1 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18446420
|
Novus Biologicals
NBP1-81353-25ul |
25ul |
285.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18248445
|
Novus Biologicals
NBP1-81353 |
0.1 mL |
500.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
YTHDC1 Polyclonal specifically detects YTHDC1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spezifikation
| YTHDC1 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| KIAA1966splicing factor YT521-B, Putative splicing factor YT521, YT521-B, YT521YTH domain-containing protein 1, YTH domain containing 1 | |
| YTHDC1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q96MU7 | |
| 91746 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LVSKPLSSSVSNNKRIVSTKGKSATEYKNEEYQRSERNKRLDADRKIRLSSSASREPYKNQPEKTCVRKRDPERRAKSPTPDGSERIGLEVD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts