Learn More
YTHDF2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26771-1-AP-20UL
 View more versions of this product
Product Code. 16885135
  • 145.00 € / 20µL
Description
The YTHDF2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human YTHDF2. This antibody recognizes human, mouse antigen. The YTHDF2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
YTHDF2
Polyclonal
Unconjugated
20 μL
51441
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
200 μg/mL
Rabbit
Primary
Human, Mouse